Anti-CD31 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: WM59]

Supplier: Biolegend
303113 303114
CA303113-BLEA 197.88 CAD
CA303113-BL CA303114-BL
Anti-CD31 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: WM59]
Antibodies
Pacific Blue™ anti-human CD31 [WM59]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: WM59
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR