Anti-CD197 (CCR7) Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: G043H7]

Supplier: Biolegend
353209 353210
CA353209-BLEA 234.58 CAD
CA353209-BL CA353210-BL
Anti-CD197 (CCR7) Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: G043H7]
Antibodies
Pacific Blue™ anti-human CD197 (CCR7) [G043H7]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD197 (CCR7)
Clonality: Monoclonal
Clone: G043H7
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR