Anti-IL-17A Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TC11-18H10.1]

Supplier: Biolegend
506907 506908
CA506907-BLEA 197.88 CAD
CA506907-BL CA506908-BL
Anti-IL-17A Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TC11-18H10.1]
Antibodies
FITC anti-mouse IL-17A [TC11-18H10.1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 500 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-17A
Clonality: Monoclonal
Clone: TC11-18H10.1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR