Anti-CD53 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: OX-79]

Supplier: Biolegend
124705 124706
CA124705-BLEA 178.73 CAD
CA124705-BL CA124706-BL
Anti-CD53 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: OX-79]
Antibodies
FITC anti-mouse CD53 [OX-79]; Isotype: Rat IgM, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD53
Clonality: Monoclonal
Clone: OX-79
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgM, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR