Anti-IgG2b, k Isotype Ctrl Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: RTK4530]

Supplier: Biolegend
400641 400642
CA400641-BLEA 307.99 CAD
CA400641-BL CA10761-184
Anti-IgG2b, k Isotype Ctrl Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: RTK4530]
Antibodies
Brilliant Violet 570™ Rat IgG2b, κ Isotype Ctrl [RTK4530]; Isotype: Rat IgG2b, κ; Reactivity: Rat; Apps: FC, ICFC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG2b, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK4530
Conjugation: Brilliant Violet® 570
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR