Anti-Ly-6C Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: HK1.4]

Supplier: Biolegend
128029 128030
CA128029-BLEA 325.54 CAD
CA128029-BL CA10757-096
Anti-Ly-6C Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: HK1.4]
Antibodies
Brilliant Violet 570™ anti-mouse Ly-6C [HK1.4]; Isotype: Rat IgG2c, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunofluorescent staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Order Now

SPECIFICATIONS

Antigen symbol Ly-6C Ly-6C
Environmentally Preferable
Antigen synonyms LY6C|locus C|Lymphocyte antigen 6 complex LY6C1|locus C|Lymphocyte antigen 6 complex
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Conjugation Brilliant Violet® 570 Brilliant Violet® 570
Clone HK1.4 HK1.4
Reactivity Mouse Mouse
Host Rat Rat
Gene ID 17067 17067
Isotype IgG2c kappa IgG2c kappa
Flow cytometry Yes
Immunogen L3 cloned CTL cells L3 cloned CTL cells
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA). Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
Purification The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 570™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 570™ and unconjugated antibody. The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 570™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 570™ and unconjugated antibody.
Concentration 0.20 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze. The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR