Anti-CD45R Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: RA3-6B2]

Supplier: Biolegend
103239 103240 103251
CA103239-BLEA 263.31 CAD
CA103239-BL CA103240-BL CA10756-836
Anti-CD45R Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: RA3-6B2]
Antibodies
Brilliant Violet 421™ anti-mouse/human CD45R/B220 [RA3-6B2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cat (Feline); Apps: FC, IHC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. For immunohistochemistry, a concentration range of 2.5 - 10 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45R
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;Human;Cross-Reactivity: Cat (Feline)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR