Anti-TNF-a Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: MP6-XT22]

Supplier: Biolegend
506327 506328
CA506327-BLEA 307.99 CAD
CA506327-BL CA10761-960
Anti-TNF-a Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: MP6-XT22]
Antibodies
Brilliant Violet 421™ anti-mouse TNF-α [MP6-XT22]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 50 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For flow cytometric staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TNF-a
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR