Anti-TER-119/Erythroid Cells Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: TER-119]
Supplier: Biolegend
116233 116234
CA116233-BLEA 307.99 CAD
CA116233-BL CA10756-976
Anti-TER-119/Erythroid Cells Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: TER-119]
Antibodies
Brilliant Violet 421™ anti-mouse TER-119/Erythroid Cells [TER-119]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. For flow cytometric staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance forMore Product Information