Anti-IL-4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 11B11]
Supplier: Biolegend
504119 504120 504127
CA504119-BLEA 307.99 CAD
CA504119-BL CA10761-368 CA10761-632
Anti-IL-4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 11B11]
Antibodies
Brilliant Violet 421™ anti-mouse IL-4 [11B11]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 50 μg
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunofluorescent staining using μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for eachMore Product Information