Anti-F4/80 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: BM8]

Supplier: Biolegend
123131 123132 123137
CA123131-BLEA 325.54 CAD
CA123131-BL CA123132-BL CA10757-016
Anti-F4/80 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: BM8]
Antibodies
Brilliant Violet 421™ anti-mouse F4/80 [BM8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunofluorescent staining using μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. For immunohistochemical staining on frozen tissue sections, the suggested use of this reagent is 2.0 μg/ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: F4/80
Clonality: Monoclonal
Clone: BM8
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR