Anti-CD8a Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 53-6.7]

Supplier: Biolegend
100737 100738 100753
CA100737-BLEA 263.31 CAD
CA100737-BL CA100738-BL CA10756-704
Anti-CD8a Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 53-6.7]
Antibodies
Brilliant Violet 421™ anti-mouse CD8a [53-6.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunofluorescent staining using μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR