Anti-CD45 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 30-F11]

Supplier: Biolegend
103133 103134
CA103133-BLEA 290.44 CAD
CA103133-BL CA10759-124
Anti-CD45 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 30-F11]
Antibodies
Brilliant Violet 421™ anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunofluorescent staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 30-F11
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR