Anti-CD25 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: PC61]

Supplier: Biolegend
102033 102034 102043
CA102033-BLEA 298.41 CAD
CA102033-BL CA102034-BL CA10756-770
Anti-CD25 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: PC61]
Antibodies
Brilliant Violet 421™ anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. For immunofluorescent staining using μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: PC61
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG1, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR