Anti-CD196 (CCR6) Armenian Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: 29-2L17]

Supplier: Biolegend
129817 129818
CA129817-BLEA 343.1 CAD
CA129817-BL CA129818-BL
Anti-CD196 (CCR6) Armenian Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: 29-2L17]
Antibodies
Brilliant Violet 421™ anti-mouse CD196 (CCR6) [29-2L17]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD196 (CCR6)
Clonality: Monoclonal
Clone: 29-2L17
Conjugation: Brilliant Violet® 421
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR