Anti-CD11c Armenian Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: N418]

Supplier: Biolegend
117329 117330 117343
CA117329-BLEA 280.86 CAD
CA117329-BL CA117330-BL CA10759-222
Anti-CD11c Armenian Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: N418]
Antibodies
Brilliant Violet 421™ anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunofluorescent staining using μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation: Brilliant Violet® 421
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR