Anti-CD117 (c-kit) Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 2B8]

Supplier: Biolegend
105827 105828
CA105827-BLEA 263.31 CAD
CA105827-BL CA105828-BL
Anti-CD117 (c-kit) Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 2B8]
Antibodies
Brilliant Violet 421™ anti-mouse CD117 (c-Kit) [2B8]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD117 (c-kit)
Clonality: Monoclonal
Clone: 2B8
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR