Anti-CD103 Armenian Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: 2E7]

Supplier: Biolegend
121421 121422
CA121421-BLEA 325.54 CAD
CA121421-BL CA10756-994
Anti-CD103 Armenian Hamster Monoclonal Antibody (Brilliant Violet® 421) [clone: 2E7]
Antibodies
Brilliant Violet 421™ anti-mouse CD103 [2E7]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For flow cytometric staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: 2E7
Conjugation: Brilliant Violet® 421
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR