Anti-HLA-DR Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: L243]

Supplier: Biolegend
307635 307636
CA307635-BLEA 290.44 CAD
CA307635-BL CA10757-966
Anti-HLA-DR Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: L243]
Antibodies
Brilliant Violet 421™ anti-human HLA-DR [L243]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Dog (Canine), Marmoset, Rhesus, Squirrel Monkey, Tamarin; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: L243
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Cynomolgus;Dog (Canine);Marmoset;Rhesus;Squirrel Monkey;Tamarin
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR