Anti-CD83 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HB15e]

Supplier: Biolegend
305323 305324
CA305323-BLEA 303.2 CAD
CA305323-BL CA10757-898
Anti-CD83 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HB15e]
Antibodies
Brilliant Violet 421™ anti-human CD83 [HB15e]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: HB15e
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Pigtailed Macaque;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR