Anti-CD45RA Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HI100]

Supplier: Biolegend
304129 304130
CA304129-BLEA 280.86 CAD
CA304129-BL CA304130-BL
Anti-CD45RA Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HI100]
Antibodies
Brilliant Violet 421™ anti-human CD45RA [HI100]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, IHC-P; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. For immunohistochemistry, a concentration range of 5.0 - 10.0 μg/ml is suggested. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: HI100
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR