Anti-CD25 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: BC96]

Supplier: Biolegend
302629 302630
CA302629-BLEA 295.22 CAD
CA302629-BL CA302630-BL
Anti-CD25 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: BC96]
Antibodies
Brilliant Violet 421™ anti-human CD25 [BC96]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC, IF; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: BC96
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Baboon;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR