Anti-CD158e1 (KIR3DL1, NKB1) Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: DX9]

Supplier: Biolegend
312713 312714
CA312713-BLEA 307.99 CAD
CA312713-BL CA10757-590
Anti-CD158e1 (KIR3DL1, NKB1) Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: DX9]
Antibodies
Brilliant Violet 421™ anti-human CD158e1 (KIR3DL1, NKB1) [DX9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD158e1 (KIR3DL1, NKB1)
Clonality: Monoclonal
Clone: DX9
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR