Anti-CD137 (4-1BB) Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 4B4-1]

Supplier: Biolegend
309819 309820
CA309819-BLEA 303.2 CAD
CA309819-BL CA10757-996
Anti-CD137 (4-1BB) Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 4B4-1]
Antibodies
Brilliant Violet 421™ anti-human CD137 (4-1BB) [4B4-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD137 (4-1BB)
Clonality: Monoclonal
Clone: 4B4-1
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR