Anti-CD117 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 104D2]

Supplier: Biolegend
313215 313216
CA313215-BLEA 325.54 CAD
CA313215-BL CA313216-BL
Anti-CD117 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 104D2]
Antibodies
Brilliant Violet 421™ anti-human CD117 (c-kit) [104D2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Cattle (Bovine, Cow); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD117
Clonality: Monoclonal
Clone: 104D2
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus;Cattle (Bovine;Cow)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR