Anti-C5a/C5a(desArg)/C5 Mouse Monoclonal Antibody (Biotin) [clone: G25/2]

Supplier: Biolegend
518305 518306
CA518305-BLEA 178.73 CAD
CA518305-BL CA518306-BL
Anti-C5a/C5a(desArg)/C5 Mouse Monoclonal Antibody (Biotin) [clone: G25/2]
Antibodies
Biotin anti-human C5a/C5a(desArg)/C5 [G25/2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Detection, WB, IP; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 0.25-1 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of C5a recombinant protein ranging from 15.6 to 1000 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: C5a/C5a(desArg)/C5
Clonality: Monoclonal
Clone: G25/2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR