Anti-CD3 Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: 17A2]

Supplier: Biolegend
100221 100222
CA100221-BLEA 167.56 CAD
CA100221-BL CA100222-BL
Anti-CD3 Rat Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: 17A2]
Antibodies
APC/Cy7 anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 17A2
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR