Anti-T-bet Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 4B10]

Supplier: Biolegend
644813 644814
CA644813-BLEA 331.93 CAD
CA644813-BL CA644814-BL
Anti-T-bet Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 4B10]
Antibodies
APC anti-T-bet [4B10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our True-Nuclear™ Transcription Factor Staining Protocol. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: T-bet
Clonality: Monoclonal
Clone: 4B10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR