Anti-TNF-a Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP6-XT22]

Supplier: Biolegend
506307 506308
CA506307-BLEA 162.77 CAD
CA506307-BL CA506308-BL
Anti-TNF-a Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP6-XT22]
Antibodies
APC anti-mouse TNF-α [MP6-XT22]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TNF-a
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR