Anti-IL-12 p40 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: C15.6]

Supplier: Biolegend
505205 505206
CA505205-BLEA 180.33 CAD
CA505205-BL CA505206-BL
Anti-IL-12 p40 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: C15.6]
Antibodies
APC anti-mouse IL-12/IL-23 p40 [C15.6]; Isotype: Rat IgG1, κ; Reactivity: Mouse, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-12 p40
Clonality: Monoclonal
Clone: C15.6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse;IL-12/IL-23 p40 subunit (monomer;homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR