Anti-I-Ab Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: AF6-120.1]

Supplier: Biolegend
116417 116418
CA116417-BLEA 151.6 CAD
CA116417-BL CA116418-BL
Anti-I-Ab Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: AF6-120.1]
Antibodies
APC anti-mouse I-Ab [AF6-120.1]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype, Cross-reacts with H-2 k and H-2 u haplotypes. Does not react with other haplotypes (e.g., d, f, q, r, s).; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: I-Ab
Clonality: Monoclonal
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR