Anti-CD54 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: YN1/1.7.4]

Supplier: Biolegend
116119 116120
CA116119-BLEA 143.62 CAD
CA116119-BL CA116120-BL
Anti-CD54 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: YN1/1.7.4]
Antibodies
APC anti-mouse CD54 [YN1/1.7.4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD54
Clonality: Monoclonal
Clone: YN1/1.7.4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR