Anti-CD115 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: AFS98]

Supplier: Biolegend
135509 135510
CA135509-BLEA 157.98 CAD
CA135509-BL CA135510-BL
Anti-CD115 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: AFS98]
Antibodies
APC anti-mouse CD115 (CSF-1R) [AFS98]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD115
Clonality: Monoclonal
Clone: AFS98
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR