Anti-IL-4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP4-25D2]

Supplier: Biolegend
500811 500812
CA500811-BLEA 157.98 CAD
CA500811-BL CA500812-BL
Anti-IL-4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP4-25D2]
Antibodies
APC anti-human IL-4 [MP4-25D2]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Swine (Pig, Porcine), Rhesus; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Human;Cross-Reactivity: Swine (Pig;Porcine);Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR