Anti-CD54 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HA58]

Supplier: Biolegend
353111 353112
CA353111-BLEA 201.07 CAD
CA353111-BL CA353112-BL
Anti-CD54 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HA58]
Antibodies
APC anti-human CD54 [HA58]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD54
Clonality: Monoclonal
Clone: HA58
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR