Anti-CD31 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: WM59]

Supplier: Biolegend
303115 303116
CA303115-BLEA 180.33 CAD
CA303115-BL CA303116-BL
Anti-CD31 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: WM59]
Antibodies
APC anti-human CD31 [WM59]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD31
Environmentally Preferable
Antigen synonyms PECAM-1|EndoCAM
Antibody type Primary
Clonality Monoclonal
Conjugation APC (Allophycocyanin)
Clone WM59
Reactivity Baboon,African green monkey,Cynomolgus monkey,Rhesus monkey,Human
Host Mouse
Gene ID 5175
Isotype IgG1 kappa
Flow cytometry Yes
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Purification The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR