Anti-CD158d Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: mAb 33 (33)]

Supplier: Biolegend
347007 347008
CA347007-BLEA 245.75 CAD
CA347007-BL CA347008-BL
Anti-CD158d Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: mAb 33 (33)]
Antibodies
APC anti-human CD158d (KIR2DL4) [mAb 33 (33)]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD158d
Clonality: Monoclonal
Clone: mAb 33 (33)
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR