Anti-CD48 Armenian Hamster Monoclonal Antibody (Alexa Fluor® 700) [clone: HM48-1]

Supplier: Biolegend
103425 103426
CA103425-BLEA 150.01 CAD
CA103425-BL CA103426-BL
Anti-CD48 Armenian Hamster Monoclonal Antibody (Alexa Fluor® 700) [clone: HM48-1]
Antibodies
Alexa Fluor® 700 anti-mouse CD48 [HM48-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: HM48-1
Conjugation: Alexa Fluor® 700
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR