Anti-CD38 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: HIT2]

Supplier: Biolegend
303523 303524
CA303523-BLEA 197.88 CAD
CA303523-BL CA303524-BL
Anti-CD38 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: HIT2]
Antibodies
Alexa Fluor® 700 anti-human CD38 [HIT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Horse, Cow (Bovine); Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Horse;Cow (Bovine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR