Anti-CD144 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: BV13]

Supplier: Biolegend
138005 138006
CA138005-BLEA 280.86 CAD
CA138005-BL CA138006-BL
Anti-CD144 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: BV13]
Antibodies
Alexa Fluor® 647 anti-mouse CD144 (VE-cadherin) [BV13]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD144
Clonality: Monoclonal
Clone: BV13
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR