Anti-IL-17A Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: BL168]

Supplier: Biolegend
512309 512310
CA512309-BLEA 263.31 CAD
CA512309-BL CA512310-BL
Anti-IL-17A Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: BL168]
Antibodies
Alexa Fluor® 647 anti-human IL-17A [BL168]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-17A
Clonality: Monoclonal
Clone: BL168
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR