Anti-FOXP3 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 206D]

Supplier: Biolegend
320113 320114
CA320113-BLEA 263.31 CAD
CA320113-BL CA320114-BL
Anti-FOXP3 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 206D]
Antibodies
Alexa Fluor® 647 anti-human FOXP3 [206D]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Pigtailed Macaque; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular flow cytometry using our True-Nuclear™ Transcription Factor Staining Protocol. For flow cytometric staining, the suggested use of this reagent is 5 μl per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: FOXP3
Clonality: Monoclonal
Clone: 206D
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR