Anti-CD305 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: NKTA255]

Supplier: Biolegend
342801 342802
CA342801-BLEA 234.58 CAD
CA342801-BL CA342802-BL
Anti-CD305 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: NKTA255]
Antibodies
Alexa Fluor® 647 anti-human CD305 (LAIR1) [NKTA255]; Isotype: Mouse IgG1; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

* Alexa Fluor® 647 has a maximum emission of 668 nm when it is excited at 633nm / 635nm.

Type: Primary
Antigen: CD305
Clonality: Monoclonal
Clone: NKTA255
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR