Anti-Bcl-2 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: BCL/10C4]

Supplier: Biolegend
633509 633510
CA633509-BLEA 205.86 CAD
CA633509-BL CA633510-BL
Anti-Bcl-2 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: BCL/10C4]
Antibodies
Alexa Fluor® 647 anti-Bcl-2 [BCL/10C4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Bcl-2
Clonality: Monoclonal
Clone: BCL/10C4
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR