Anti-TNF-a Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: MP6-XT22]

Supplier: Biolegend
506313 506315
CA506313-BLEA 427.67 CAD
CA506313-BL CA506315-BL
Anti-TNF-a Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: MP6-XT22]
Antibodies
Alexa Fluor® 488 anti-mouse TNF-α [MP6-XT22]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

* Alexa Fluor® 488 has a maximum emission of 519 nm when it is excited at 488 nm.

Type: Primary
Antigen: TNF-a
Clonality: Monoclonal
Clone: MP6-XT22
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol TNF-a
Environmentally Preferable
Antigen synonyms Differentiation inducing factor (DIF)|Necrosin|TNFSF2|TNFA|Cachectin|Tumor necrosis factor-α|Macrophage cytotoxic factor (MCF)
Antibody type Primary
Clonality Monoclonal
Conjugation Alexa Fluor® 488
Clone MP6-XT22
Reactivity Mouse
Host Rat
Gene ID 21926
Isotype IgG1 kappa
Flow cytometry Yes
Immunogen E. coli-expressed, recombinant mouse TNF-α
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 488 under optimal conditions.
Concentration 0.50 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR