Anti-IFN-g Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: XMG1.2]

Supplier: Biolegend
505813 505815
CA505813-BLEA 402.14 CAD
CA505813-BL CA505815-BL
Anti-IFN-g Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: XMG1.2]
Antibodies
Alexa Fluor® 488 anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFN-g
Clonality: Monoclonal
Clone: XMG1.2
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR