Anti-CD1c Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: L161]

Supplier: Biolegend
331521 331522
CA331521-BLEA 217.03 CAD
CA331521-BL CA331522-BL
Anti-CD1c Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: L161]
Antibodies
Alexa Fluor® 488 anti-human CD1c [L161]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD1c
Clonality: Monoclonal
Clone: L161
Conjugation: Alexa Fluor® 488
Epitope:
Host:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR