Anti-Bcl-2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: BCL/10C4]

Supplier: Biolegend
633505 633506
CA633505-BLEA 197.88 CAD
CA633505-BL CA633506-BL
Anti-Bcl-2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: BCL/10C4]
Antibodies
Alexa Fluor® 488 anti-Bcl-2 [BCL/10C4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: ICFC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Bcl-2
Clonality: Monoclonal
Clone: BCL/10C4
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR