Anti-IgG Donkey Polyclonal Antibody (CF™ 640R)

Supplier: Biotium

20179 20179-1
97065-564EA 411.57 CAD
97065-564 97065-566
Anti-IgG Donkey Polyclonal Antibody (CF™ 640R)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 640R
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR