Anti-IgG Rabbit Polyclonal Antibody (CF™ 640R)

Supplier: Biotium

20201 20201-1
97065-624EA 360.95 CAD
97065-624 97065-626
Anti-IgG Rabbit Polyclonal Antibody (CF™ 640R)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 640R
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR